missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Endoglin/CD105 Recombinant Protein

For use in cell or tissue culture applications.

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2022
Molecular Weight 61 kDa
Name Endoglin/CD105
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1097EN025CF R&D Systems
RDS1097EN025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
1097EN025 R&D Systems
RDS1097EN025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
Specifications

Description

  • Source: Mouse myeloma cell line, NS0-derived, Glu26-Gly586
  • Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
  • Molecular Weight: 61 kDa
  • Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
  • Entrez Gene ID: 2022
  • Regulatory Status: RUO
Description & Specifications

Specifications

Unlabeled
61 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
2022
Endoglin/CD105
25μg
Human