Please login to your online account to display your discounted pricing

R&D Systems Human EG-VEGF/PK1 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 84432
Molecular Weight 10.5 kDa
Name EG-VEGF/PK1
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1209EV025

 
r&d systems
RDS1209EV025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87

1209EV025CF

 
r&d systems
RDS1209EV025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 84432
Molecular Weight 10.5 kDa
Name EG-VEGF/PK1
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: Mouse myeloma cell line, NS0-derived, Ala20-Phe105, with a C-terminal 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 10.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 84432
Regulatory Status: RUO