missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human DLL4 Recombinant Protein

$415.83 - $415.83

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 54567
Molecular Weight 55.6 kDa
Name DLL4
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1506D4050 R&D Systems
RDS1506D4050
Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein. Each for $415.83
1506D4050CF R&D Systems
RDS1506D4050CF
Lyophilized from a 0.2μm filtered solution in PBS and NaCl. Each for $415.83
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ser27-Pro524, with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 55.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
Entrez Gene ID: 54567
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
55.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
54567
DLL4
50μg
Human