Please login to your online account to display your discounted pricing

R&D Systems Human Dkk-4 Recombinant Protein


Manufacturer: r&d systems  RDS1269DK010

 View more versions of this product

Catalog No. 1269DK010

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 24.4 kDa
Name Dkk-4
Gene ID (Entrez) 27121
Formulation Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl with BSA as a carrier protein.
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Source Human
Quantity 10μg
Conjugate Unlabeled
Recombinant Yes