Please login to your online account to display your discounted pricing

R&D Systems Human Dkk-1 Recombinant Protein

$415.83 - $415.83

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 22943
Molecular Weight 25.8 kDa
Name Dkk-1
Purity 95%, by SDS-PAGE with silver staining.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

5439DK010

 
r&d systems
RDS5439DK010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $415.83

5439DK010CF

 
r&d systems
RDS5439DK010CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $415.83
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 22943
Molecular Weight 25.8 kDa
Name Dkk-1
Purity 95%, by SDS-PAGE with silver staining.
Quantity 10μg
Recombinant Yes
Source Human

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Thr32-His266
Purity: 95%, by SDS-PAGE with silver staining.
Molecular Weight: 25.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 22943
Regulatory Status: RUO