missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CXCL9/MIG Recombinant Protein

$259.40 - $1,041.55

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 4283
Molecular Weight 11.7 kDa
Name CXCL9/MIG
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
392MG010 R&D Systems
RDS392MG010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 10μg Each for $259.40
392MG010CF R&D Systems
RDS392MG010CF
Lyophilized from a 0.2μm filtered solution in PBS. 10μg Each for $259.40
392MG050 R&D Systems
RDS392MG050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 50μg Each for $1,041.55
392MG050CF R&D Systems
RDS392MG050CF
Lyophilized from a 0.2μm filtered solution in PBS. 50μg Each for $1,041.55
Specifications

Description

Source: E. coli-derived, Thr23-Thr125
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 11.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 4283
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
11.7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
4283
CXCL9/MIG
Yes