missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CXCL8/IL-8 Recombinant Protein

$194.23 - $520.12

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3576
Molecular Weight 8 kDa
Name CXCL8/IL-8
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
208IL010 R&D Systems
RDS208IL010
Lyophilized from a 0.2μm filtered solution in deionized water with BSA as a carrier protein. 10μg Each for $194.23
208IL010CF R&D Systems
RDS208IL010CF
Lyophilized from a 0.2μm filtered solution in deionized water. 10μg Each for $194.23
208IL050 R&D Systems
RDS208IL050
Lyophilized from a 0.2μm filtered solution in deionized water with BSA as a carrier protein. 50μg Each for $520.12
208IL050CF R&D Systems
RDS208IL050CF
Lyophilized from a 0.2μm filtered solution in deionized water. 50μg Each for $520.12
Specifications

Description

Source: E. coli-derived, Ser28-Ser99
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in deionized water.
Entrez Gene ID: 3576
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
3576
CXCL8/IL-8
Yes