Please login to your online account to display your discounted pricing

R&D Systems Human CXCL8/IL-8 Mucin Stalk Chimera Recombinant Protein

$292.21 - $292.21

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3576
Molecular Weight 34.8 kDa
Name CXCL8/IL-8 Mucin Stalk Chimera
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

968IL025

 
r&d systems
RDS968IL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $292.21

968IL025CF

 
r&d systems
RDS968IL025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $292.21
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3576
Molecular Weight 34.8 kDa
Name CXCL8/IL-8 Mucin Stalk Chimera
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: Mouse myeloma cell line, NS0-derived, Ala23-Ser99 & Ser28-Ser99
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 34.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3576
Regulatory Status: RUO