missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CXCL7/NAP-2 Recombinant Protein

$298.51 - $520.12

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 5473
Molecular Weight 7.6 kDa
Name CXCL7/NAP-2
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
393NP010 R&D Systems
RDS393NP010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 10μg Each for $298.51
393NP010CF R&D Systems
RDS393NP010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 10μg Each for $298.51
393NP050 R&D Systems
RDS393NP050
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 50μg Each for $520.12
393NP050CF R&D Systems
RDS393NP050CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 50μg Each for $520.12
Specifications

Description

Source: E. coli-derived, Ala59-Asp128
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 5473
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
7.6 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
5473
CXCL7/NAP-2
Yes