missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CXCL6/GCP-2 Recombinant Protein

$415.83 - $415.83

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6372
Molecular Weight 8 kDa
Name CXCL6/GCP-2
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
333GC025 R&D Systems
RDS333GC025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $415.83
333GC025CF R&D Systems
RDS333GC025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $415.83
Specifications

Description

Source: E. coli-derived, Val40-Asn114
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6372
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
6372
CXCL6/GCP-2
25μg
Human