Please login to your online account to display your discounted pricing

R&D Systems Human CXCL5/ENA-78 Recombinant Protein

$427.46 - $427.46

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6374
Molecular Weight 8.4 kDa
Name CXCL5/ENA-78
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

254XB025

 
r&d systems
RDS254XB025
Lyophilized from a 0.2mm filtered solution in PBS containing BSA as carrier protein Each for $427.46

254XB025CF

 
r&d systems
RDS254XB025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $427.46
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6374
Molecular Weight 8.4 kDa
Name CXCL5/ENA-78
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: E. coli-derived, Ala37-Asn114
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 6374
Regulatory Status: RUO