missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CXCL5/ENA-74 Recombinant Protein

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6374
Molecular Weight 8.0 kDa
Name CXCL5/ENA-74
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
651NA025 R&D Systems
RDS651NA025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $298.51
651NA025CF R&D Systems
RDS651NA025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $298.51
Specifications

Description

Source: E. coli-derived, Ala41-Asn114
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.0 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6374
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
8.0 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
6374
CXCL5/ENA-74
25μg
Human