Please login to your online account to display your discounted pricing

R&D Systems Human CXCL5/ENA-70 Recombinant Protein

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6374
Molecular Weight 7.7 kDa
Name CXCL5/ENA-70
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

649EN025CF

 
r&d systems
RDS649EN025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $298.51

649EN025

 
r&d systems
RDS649EN025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $298.51
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6374
Molecular Weight 7.7 kDa
Name CXCL5/ENA-70
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: E. coli-derived, Arg45-Asn114
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6374
Regulatory Status: RUO