missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CXCL4/PF4 Recombinant Protein

$328.11 - $404.12

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 5196
Molecular Weight 7.8 kDa
Name CXCL4/PF4
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Purity Price Quantity  
Catalog Number Mfr. No. Formulation Purity Price Quantity  
795P4025 R&D Systems
RDS795P4025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. Each for $404.12
795P4025CF R&D Systems
RDS795P4025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. Each for $404.12
7989P4025CF R&D Systems
RDS7989P4025CF
Lyophilized from a 0.2μm filtered solution in PBS. 95%, by SDS-PAGE with silver staining. Each for $328.11
7989P4025 R&D Systems
RDS7989P4025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 95%, by SDS-PAGE with silver staining. Each for $328.11
Specifications

Description

Source: Human embryonic kidney cell, HEK293-derived, Glu32-Ser101
Purity: 95%, by SDS-PAGE with silver staining.
Molecular Weight: 7.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 5196
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
7.8 kDa
25μg
Human
5196
CXCL4/PF4
Yes