missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CXCL3/GRO gamma/CINC-2/DCIP-1 Recombinant Protein

$272.44 - $520.12

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2921
Molecular Weight 8 kDa
Name CXCL3/GRO gamma/CINC-2/DCIP-1
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
277GG010 R&D Systems
RDS277GG010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 10μg Each for $272.44
277GG010CF R&D Systems
RDS277GG010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 10μg Each for $272.44
277GG050 R&D Systems
RDS277GG050
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 50μg Each for $520.12
277GG050CF R&D Systems
RDS277GG050CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 50μg Each for $520.12
Specifications

Description

Source: E. coli-derived, Ala35-Asn107
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 2921
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
2921
CXCL3/GRO gamma/CINC-2/DCIP-1
Yes