missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CXCL16 Extracellular Domain Recombinant Protein, Asn49-Thr224

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 58191
Molecular Weight 20 kDa
Name CXCL16 Extracellular Domain
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1164CX025 R&D Systems
RDS1164CX025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $298.51
1164CX025CF R&D Systems
RDS1164CX025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $298.51
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Asn49-Thr224, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 58191
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
20 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
58191
CXCL16 Extracellular Domain
25μg
Human