Please login to your online account to display your discounted pricing

R&D Systems Human CXCL16 Extracellular Domain Recombinant Protein, Asn49-Thr224


Manufacturer: r&d systems  RDS1164CX025CF

 View more versions of this product

Catalog No. 1164CX025CF

  • / Each
 

Description & Specifications

Specifications

Name CXCL16 Extracellular Domain
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight 20 kDa
Recombinant Yes
Quantity 25μg
Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 58191
Source Human