missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CXCL12/SDF-1 gamma Recombinant Protein

$378.79 - $378.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6387
Molecular Weight 11.6 kDa
Name CXCL12/SDF-1 gamma
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
6448SD025 R&D Systems
RDS6448SD025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $378.79
6448SD025CF R&D Systems
RDS6448SD025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $378.79
Specifications

Description

Source: E. coli-derived, Gly21-Asn119
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 11.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 6387
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
11.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
6387
CXCL12/SDF-1 gamma
25μg
Human