Please login to your online account to display your discounted pricing

R&D Systems Human CXCL10/IP-10/CRG-2 Recombinant Protein

$151.85 - $470.86

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3627
Molecular Weight 8.7 kDa
Name CXCL10/IP-10/CRG-2
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

266IP010

 
r&d systems
RDS266IP010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 10μg Each for $151.85

266IP050

 
r&d systems
RDS266IP050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 50μg Each for $470.86

266IP050CF

 
r&d systems
RDS266IP050CF
Lyophilized from a 0.2μm filtered solution in PBS. 50μg Each for $470.86

266IP010CF

 
r&d systems
RDS266IP010CF
Lyophilized from a 0.2μm filtered solution in PBS. 10μg Each for $151.85
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3627
Molecular Weight 8.7 kDa
Name CXCL10/IP-10/CRG-2
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Human

Source: E. coli-derived, Val22-Pro98, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3627
Regulatory Status: RUO