missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CXCL1/GRO alpha/KC/CINC-1 Recombinant Protein

$259.40 - $780.83

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2919
Molecular Weight 7.9 kDa
Name CXCL1/GRO alpha/KC/CINC-1
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
275GR010 R&D Systems
RDS275GR010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 10μg Each for $259.40
275GR050 R&D Systems
RDS275GR050
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 50μg Each for $780.83
275GR010CF R&D Systems
RDS275GR010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 10μg Each for $259.40
275GR050CF R&D Systems
RDS275GR050CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 50μg Each for $780.83
Specifications

Description

Source: E. coli-derived, Ala35-Asn107
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 2919
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
7.9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
2919
CXCL1/GRO alpha/KC/CINC-1
Yes