missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CX3CL1/Fractalkine Full Length Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6376
Molecular Weight 35 kDa
Name CX3CL1/Fractalkine Full Length
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
365FR025 R&D Systems
RDS365FR025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
365FR025CF R&D Systems
RDS365FR025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Gln25-Arg339, with a C-terminal 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 35 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 6376
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
35 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
6376
CX3CL1/Fractalkine Full Length
25μg
Human