Please login to your online account to display your discounted pricing

R&D Systems Human CX3CL1/Fractalkine Chemokine Domain Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6376
Molecular Weight 8.5 kDa
Name CX3CL1/Fractalkine Chemokine Domain
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

362CX025

 
r&d systems
RDS362CX025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $419.81

362CX025CF

 
r&d systems
RDS362CX025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6376
Molecular Weight 8.5 kDa
Name CX3CL1/Fractalkine Chemokine Domain
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: E. coli-derived, Gln25-Gly100
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6376
Regulatory Status: RUO