missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CTLA-4 Fc Chimera Recombinant Protein

$467.46 - $467.46

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 1493
Molecular Weight 40 kDa (monomer)
Name CTLA-4 Fc Chimera
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
325CT200 R&D Systems
RDS325CT200
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $467.46
325CT200CF R&D Systems
RDS325CT200CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $467.46
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ala37-Phe162, with a C-terminal 6-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 40 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 1493
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
40 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
1493
CTLA-4 Fc Chimera
200μg
Human