missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human COCO Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 199699
Molecular Weight 18.2 kDa (monomer)
Name COCO
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3047CC050 R&D Systems
RDS3047CC050
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $428.87
3047CC050CF R&D Systems
RDS3047CC050CF
Supplied as a 0.2μm filtered solution in Acetonitrile and TFA. Each for $428.87
Specifications

Description

Source: E. coli-derived, Arg23-Ala189, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.2 kDa (monomer)
Formulation: Supplied as a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 199699
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18.2 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
199699
COCO
50μg
Human