missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CNTF R alpha Recombinant Protein

$467.98 - $467.98

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 1271
Molecular Weight 36 kDa
Name CNTF R alpha
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
303CR050 R&D Systems
RDS303CR050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $467.98
303CR050CF R&D Systems
RDS303CR050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $467.98
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Gln23-Pro346
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 36 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 1271
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
36 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
1271
CNTF R alpha
50μg
Human