Please login to your online account to display your discounted pricing

R&D Systems Human CLC Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 23529
Molecular Weight 22.4 kDa
Name CLC
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

962CL050

 
r&d systems
RDS962CL050
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $428.87

962CL050CF

 
r&d systems
RDS962CL050CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 23529
Molecular Weight 22.4 kDa
Name CLC
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
Recombinant Yes
Source Human

Source: E. coli-derived, Leu28-Phe225, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 23529
Regulatory Status: RUO