missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CLC/CNTF R alpha Chimera Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Molecular Weight 61 kDa
Name CLC/CNTF R alpha Chimera
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2415CR025 R&D Systems
RDS2415CR025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
2415CR025CF R&D Systems
RDS2415CR025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Leu28-Phe225, with C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 61 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
CLC/CNTF R alpha Chimera
25μg
Human
61 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes