missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Chemerin Recombinant Protein

$415.83 - $415.83

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 5919
Molecular Weight 16 kDa
Name Chemerin
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2324CM025 R&D Systems
RDS2324CM025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $415.83
2324CM025CF R&D Systems
RDS2324CM025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $415.83
Specifications

Description

Source: E. coli-derived, Glu21-Ser157, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 5919
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
16 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
5919
Chemerin
25μg
Human