missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CD84/SLAMF5 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 8832
Molecular Weight 23.6 kDa
Name CD84/SLAMF5
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1855CD050 R&D Systems
RDS1855CD050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
1855CD050CF R&D Systems
RDS1855CD050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Lys22-Arg220, with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 23.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 8832
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
23.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
8832
CD84/SLAMF5
50μg
Human