missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CD58/LFA-3 Recombinant Protein

$481.01 - $481.01

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 965
Molecular Weight 22.8 kDa
Name CD58/LFA-3
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1689CD050 R&D Systems
RDS1689CD050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $481.01
1689CD050CF R&D Systems
RDS1689CD050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $481.01
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Phe29-Arg215, with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 965
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
22.8 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
965
CD58/LFA-3
50μg
Human