missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CD40 Ligand/TNFSF5 Recombinant Protein

$389.76 - $389.76

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 959
Molecular Weight 21.6 kDa
Name CD40 Ligand/TNFSF5
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
6420CL025CF R&D Systems
RDS6420CL025CF
Lyophilized from a 0.2μm filtered solution in PBS and EDTA. Each for $389.76
6420CL025 R&D Systems
RDS6420CL025
Lyophilized from a 0.2μm filtered solution in PBS and EDTA with BSA as a carrier protein. Each for $389.76
Specifications

Description

Source: Human embryonic kidney cell, HEK293-derived, Met113-Leu261
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and EDTA.
Entrez Gene ID: 959
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
21.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
959
CD40 Ligand/TNFSF5
25μg
Human