missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CD40 Ligand/TNFSF5 (Histidine-tagged) Recombinant Protein

$311.55 - $311.55

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 959
Molecular Weight 17.7 kDa (monomer)
Name CD40 Ligand/TNFSF5 (Histidine-tagged)
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2706CL025 R&D Systems
RDS2706CL025
Lyophilized from a 0.2μm filtered solution in Phosphate, NaCl and EDTA with BSA as a carrier protein. Each for $311.55
2706CL025CF R&D Systems
RDS2706CL025CF
Lyophilized from a 0.2μm filtered solution in Phosphate, NaCl and EDTA. Each for $311.55
Specifications

Description

Source: E. coli-derived, Glu108-Leu261, with an N-terminal Met and 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.7 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Phosphate, NaCl and EDTA.
Entrez Gene ID: 959
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17.7 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
959
CD40 Ligand/TNFSF5 (Histidine-tagged)
25μg
Human