missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CD40 Ligand/TNFSF5 aa 108-261 Recombinant Protein

$415.83 - $415.83

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 959
Molecular Weight 16.9 kDa (monomer)
Name CD40 Ligand/TNFSF5 aa 108-261
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
6245CL050 R&D Systems
RDS6245CL050
Lyophilized from a 0.2μm filtered solution in NaH2PO4, NaCl and EDTA with BSA as a carrier protein. Each for $415.83
6245CL050CF R&D Systems
RDS6245CL050CF
Lyophilized from a 0.2μm filtered solution in NaH2PO4, NaCl and EDTA. Each for $415.83
Specifications

Description

Source: E. coli-derived, Glu108-Leu261, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16.9 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in NaH2PO4, NaCl and EDTA.
Entrez Gene ID: 959
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
16.9 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
959
CD40 Ligand/TNFSF5 aa 108-261
50μg
Human