missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CD25/IL-2 R alpha Recombinant Protein

$246.37 - $741.73

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3559
Molecular Weight 22 kDa
Name CD25/IL-2 R alpha
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
2232A005 R&D Systems
RDS2232A005
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 5μg Each for $246.37
2232A005CF R&D Systems
RDS2232A005CF
Lyophilized from a 0.2μm filtered solution in PBS. 5μg Each for $246.37
2232A025CF R&D Systems
RDS2232A025CF
Lyophilized from a 0.2μm filtered solution in PBS. 25μg Each for $741.73
2232A025 R&D Systems
RDS2232A025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 25μg Each for $741.73
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Glu22-Cys213
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3559
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
22 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
3559
CD25/IL-2 R alpha
Yes