missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CD155/PVR Recombinant Protein

$429.46 - $429.46

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 5817
Molecular Weight 35.3 kDa
Name CD155/PVR
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2530CD050 R&D Systems
RDS2530CD050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $429.46
2530CD050CF R&D Systems
RDS2530CD050CF
Supplied as a 0.2μm filtered solution in PBS. Each for $429.46
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Gly27-Asn343, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 35.3 kDa
Formulation: Supplied as a 0.2μm filtered solution in PBS.
Entrez Gene ID: 5817
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
35.3 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
5817
CD155/PVR
50μg
Human