missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CCL8/MCP-2 Recombinant Protein

$259.40 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6355
Molecular Weight 9 kDa
Name CCL8/MCP-2
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
281CP010 R&D Systems
RDS281CP010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 10μg Each for $259.40
281CP010CF R&D Systems
RDS281CP010CF
Lyophilized from a 0.2μm filtered solution in PBS. 10μg Each for $259.40
281CP050 R&D Systems
RDS281CP050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 50μg Each for $428.87
281CP050CF R&D Systems
RDS281CP050CF
Lyophilized from a 0.2μm filtered solution in PBS. 50μg Each for $428.87
Specifications

Description

Source: E. coli-derived, Gln24-Pro99
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 6355
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
6355
CCL8/MCP-2
Yes