missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CCL5/RANTES Recombinant Protein, N-terminal Met

$311.55 - $311.55

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6352
Molecular Weight 7.8 kDa
Name CCL5/RANTES
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
335RM025 R&D Systems
RDS335RM025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $311.55
335RM025CF R&D Systems
RDS335RM025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $311.55
Specifications

Description

Source: E. coli-derived, Ser24-Ser91, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6352
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
7.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
6352
CCL5/RANTES
25μg
Human