Please login to your online account to display your discounted pricing

R&D Systems Human CCL4/MIP-1 beta Recombinant Protein

$259.40 - $559.23

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6351
Name CCL4/MIP-1 beta
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
View More Specs
Products
Catalog Number Mfr. No. Formulation Molecular Weight Quantity Price Quantity    

271BME010

 
r&d systems
RDS271BME010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 7.8 kDa & 7.3 kDa 10μg Each for $259.40

271BME050

 
r&d systems
RDS271BME050
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 7.8 kDa & 7.3 kDa 50μg Each for $559.23

271BME010CF

 
r&d systems
RDS271BME010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 7.8 kDa (major) & 7.3 kDa (minor) 10μg Each for $259.40

271BME050CF

 
r&d systems
RDS271BME050CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 7.8 kDa (major) & 7.3 kDa (minor) 50μg Each for $559.23
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6351
Name CCL4/MIP-1 beta
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Human

Source: E. coli-derived, Ala24-Asn92 (Major); Pro30-Asn92 (minor)
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.8 kDa (major) & 7.3 kDa (minor)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6351
Regulatory Status: RUO