Please login to your online account to display your discounted pricing

R&D Systems Human CCL27/CTACK Recombinant Protein

$311.55 - $311.55

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 10850
Molecular Weight 10.1 kDa
Name CCL27/CTACK
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

376CT025

 
r&d systems
RDS376CT025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $311.55

376CT025CF

 
r&d systems
RDS376CT025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $311.55
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 10850
Molecular Weight 10.1 kDa
Name CCL27/CTACK
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: E. coli-derived, Phe25-Gly112
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 10.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 10850
Regulatory Status: RUO