Please login to your online account to display your discounted pricing

R&D Systems Human CCL25/TECK Recombinant Protein

$465.75 - $465.75

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6370
Molecular Weight 14.3 kDa
Name CCL25/TECK
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

334TK025

 
r&d systems
RDS334TK025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $465.75

334TK025CF

 
r&d systems
RDS334TK025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $465.75
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6370
Molecular Weight 14.3 kDa
Name CCL25/TECK
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: E. coli-derived, Gln24-Lys150, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6370
Regulatory Status: RUO