missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CCL23/MPIF-1 aa 46-120 Recombinant Protein

$264.77 - $264.77

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6368
Molecular Weight 8.6 kDa
Name CCL23/MPIF-1 aa 46-120
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
131M1025 R&D Systems
RDS131M1025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $264.77
131M1025CF R&D Systems
RDS131M1025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $264.77
Specifications

Description

Source: E. coli-derived, Arg46-Asn120
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6368
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
8.6 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
6368
CCL23/MPIF-1 aa 46-120
25μg
Human