Please login to your online account to display your discounted pricing

R&D Systems Human CCL2/JE/MCP-1 Mucin Stalk Chimera Recombinant Protein

$272.44 - $272.44

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6347
Molecular Weight 35 kDa
Name CCL2/JE/MCP-1 Mucin Stalk Chimera
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

979MC025

 
r&d systems
RDS979MC025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $272.44

979MC025CF

 
r&d systems
RDS979MC025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $272.44
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6347
Molecular Weight 35 kDa
Name CCL2/JE/MCP-1 Mucin Stalk Chimera
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: Mouse myeloma cell line, NS0-derived, Gln24-Thr99
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 35 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 6347
Regulatory Status: RUO