missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CCL19/MIP-3 beta Recombinant Protein

$404.12 - $404.12

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6363
Molecular Weight 8.8 kDa
Name CCL19/MIP-3 beta
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
361MI025 R&D Systems
RDS361MI025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $404.12
361MI025CF R&D Systems
RDS361MI025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $404.12
Specifications

Description

Source: E. coli-derived, Gly22-Ser98
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6363
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
8.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
6363
CCL19/MIP-3 beta
25μg
Human