Please login to your online account to display your discounted pricing

R&D Systems Human CCL16/HCC-4 Recombinant Protein

$272.44 - $272.44

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6360
Molecular Weight 11 kDa
Name CCL16/HCC-4
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

802HC025CF

 
r&d systems
RDS802HC025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $272.44

802HC025

 
r&d systems
RDS802HC025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $272.44
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6360
Molecular Weight 11 kDa
Name CCL16/HCC-4
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: E. coli-derived, Gln24-Gln120
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 11 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6360
Regulatory Status: RUO