missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CCL15/MIP-1 delta 92 aa Recombinant Protein

$272.44 - $272.44

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6359
Molecular Weight 10 kDa
Name CCL15/MIP-1 delta 92 aa
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
363MG025CF R&D Systems
RDS363MG025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $272.44
363MG025 R&D Systems
RDS363MG025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $272.44
Specifications

Description

Source: E. coli-derived, Gln22-Ile113
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 10 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6359
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
10 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
6359
CCL15/MIP-1 delta 92 aa
25μg
Human