missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CCL14a/HCC-1 aa 28-93 Recombinant Protein

$290.11 - $290.11

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6358
Molecular Weight 7.9 kDa
Name CCL14a/HCC-1 aa 28-93
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1578HC025 R&D Systems
RDS1578HC025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $290.11
1578HC025CF R&D Systems
RDS1578HC025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $290.11
Specifications

Description

Source: E. coli-derived, Gly28-Asn93, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 6358
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
7.9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
6358
CCL14a/HCC-1 aa 28-93
25μg
Human