missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CCL14a/HCC-1 aa 20-93 Recombinant Protein

$252.10 - $378.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6358
Molecular Weight 8.7 kDa
Name CCL14a/HCC-1 aa 20-93
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
324HC010CF R&D Systems
RDS324HC010CF
Lyophilized from a 0.2μm filtered solution in PBS. 10μg Each for $252.10
324HC010 R&D Systems
RDS324HC010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 10μg Each for $252.10
324HC050 R&D Systems
RDS324HC050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 50μg Each for $378.79
324HC050CF R&D Systems
RDS324HC050CF
Lyophilized from a 0.2μm filtered solution in PBS. 50μg Each for $378.79
Specifications

Description

Source: E. coli-derived, Thr20-Asn93
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 6358
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
8.7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
6358
CCL14a/HCC-1 aa 20-93
Yes