missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human CCL13/MCP-4 Recombinant Protein

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6357
Molecular Weight 8.6 kDa
Name CCL13/MCP-4
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
327P4025 R&D Systems
RDS327P4025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $298.51
327P4025CF R&D Systems
RDS327P4025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $298.51
Specifications

Description

Source: E. coli-derived, Gln24-Thr98
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 6357
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
8.6 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
6357
CCL13/MCP-4
25μg
Human