Please login to your online account to display your discounted pricing

R&D Systems Human CCL1/I-309/TCA-3 Recombinant Protein

$266.69 - $547.42

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6346
Molecular Weight 8.5 kDa
Name CCL1/I-309/TCA-3
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

272I010

 
r&d systems
RDS272I010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 10μg Each for $266.69

272I050

 
r&d systems
RDS272I050
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 50μg Each for $547.42

272I050CF

 
r&d systems
RDS272I050CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 50μg Each for $547.42

272I010CF

 
r&d systems
RDS272I010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 10μg Each for $266.69
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6346
Molecular Weight 8.5 kDa
Name CCL1/I-309/TCA-3
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Human

Source: E. coli-derived, Lys24-Lys96
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 6346
Regulatory Status: RUO