missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Caspase-8 Recombinant Protein

$467.46 - $467.46

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 841
Molecular Weight 18 kDa & 10 kDa
Name Caspase-8
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
705C8010CF R&D Systems
RDS705C8010CF
Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Sucrose. Each for $467.46
Specifications

Description

Source: E. coli-derived, Ser217-Asp384 (Asp285His) (p18 subunit) & Leu385-Asp479 (p10 subunit), with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa & 10 kDa
Formulation: Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Sucrose.
Entrez Gene ID: 841
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18 kDa & 10 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
841
Caspase-8
10μg
Human